Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™
Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™
Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™
Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™
Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™
Thermo Scientific Chemicals
Sodium borohydride, 12% solution in 40% aq. NaOH solution, AcroSeal™
CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA429131000100 mLAcroSeal™ Glass Bottle
Catalog number FSA429131000
Price (MYR)
514.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
514.00
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
Assay from Supplier's CofA≥99.99% (metals basis)
FormSlug, 3.175mm (0.125in) diameter.
Reduction reagent
RUO – Research Use Only