N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
Thermo Scientific Chemicals
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride, 98%
CAS: 21653-40-7 | C19H22N6O4·HCl | 434.88 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4286400101 gGlass bottle
Catalog number FSA428640010
Price (MYR)
4,808.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,808.00
Add to cart
Chemical Identifiers
CAS95-50-1
IUPAC Name1,2-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyRFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.5500 to 1.5520 (20°C, 589 nm)