Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
Thermo Scientific Chemicals
Boron trifluoride dibutyl etherate, 30-35%, (BF3), AcroSeal™
CAS: 593-04-4 | C8H18BF3O | 198.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA428141000100 mLAcroSeal™ Glass Bottle
Catalog number FSA428141000
Price (MYR)
762.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
762.00
Add to cart
Chemical Identifiers
CAS100-63-0
IUPAC Namehydrogen phenylhydrazine chloride
Molecular FormulaC6H9ClN2
InChI KeyJOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light orange
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=96.0 %
Lewis acid
RUO – Research Use Only