L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-(+)-Tartaric acid dipotassium salt hemihydrate, 97%, Thermo Scientific Chemicals
CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA427991000
Price (MYR)
353.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
353.00
Add to cart
Chemical Identifiers
CAS927-58-2
IUPAC Name4-bromobutanoyl chloride
Molecular FormulaC4H6BrClO
InChI KeyLRTRXDSAJLSRTG-UHFFFAOYSA-N
SMILESClC(=O)CCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow to light pink
Refractive index1.4910 to 1.4940 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=94 %