Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals
CAS: 23519-77-9 | C12H28O4Zr | 327.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA427931000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13453-69-5
IUPAC Nametetraboron heptahydrate dilithium
Molecular FormulaB4H14Li2O7
InChI KeyDPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<200 ppm
Iron (Fe)=<50 ppm
Appearance (Color)White
Appearance (Form)Powder
Titration with HCl>=98.5 %