DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-3-Phenyllactic acid, 97%, Thermo Scientific Chemicals
CAS: 828-01-3 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number FSA427770010
Price (MYR)
318.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Chemical Identifiers
CAS30525-89-4
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or beads
Infrared spectrumConforms
Titration with HCl>=95 %
Ash=<0.02 %