Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophorone diisocyanate, 98+%, mixture of isomers, Thermo Scientific Chemicals
CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA427600500
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-88-6
IUPAC Name2,3-dichloroprop-1-ene
Molecular FormulaC3H4Cl2
InChI KeyFALCMQXTWHPRIH-UHFFFAOYSA-N
SMILESClCC(Cl)=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4601 to 1.4621 (20°C, 589 nm)
Appearance (Color)Clear colorless to pink-brown
Appearance (Form)Liquid