Tri-n-butylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals
Tri-n-butylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals
Tri-n-butylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals
Tri-n-butylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals
Alfa Aesar
Tri-n-butylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals
CAS: 998-40-3 | C12H27P | 202.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA427308000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS10034-93-2
IUPAC Namehydrazine; sulfuric acid
Molecular FormulaH6N2O4S
InChI KeyZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILESNN.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Zinc (Zn)=<5 ppm
Magnesium (Mg)=<5 ppm
Total chloride=<20 ppm
Appearance (Form)Crystalline powder
Calcium (Ca)=<10 ppm