Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
Thermo Scientific Chemicals
Methanol-d4, for NMR, 99.8 atom% D, AcroSeal™
CAS: 811-98-3 | CH4O | 36.066 g/mol
Have Questions?
Catalog NumberQuantity
FSA426951000100 mL
Catalog number FSA426951000
Price (MYR)
3,754.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
3,754.00
Add to cart
Chemical Identifiers
CAS762-21-0
IUPAC Name1,4-diethyl but-2-ynedioate
Molecular FormulaC8H10O4
InChI KeySTRNXFOUBFLVIN-UHFFFAOYSA-N
SMILESCCOC(=O)C#CC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.4410 to 1.4430 (20°C, 589 nm)
For NMR
RUO – Research Use Only