Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
Thermo Scientific Chemicals
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™
CAS: 7789-20-0 | H2O | 20.03 g/mol
Have Questions?
Catalog NumberQuantity
FSA426931000100 mL
Catalog number FSA426931000
Price (MYR)
1,263.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
1,263.00
Add to cart
Chemical Identifiers
CAS919-30-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
CommentMay contain very small amount of sediment/precipitate
Refractive Index1.4190-1.4230 @20?C
View more
For NMR
RUO – Research Use Only