1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
CAS: 32387-21-6 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA426480010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14040-11-0
IUPAC Nametungsten hexacarbonyl
Molecular FormulaC6O6W
InChI KeyFQNHWXHRAUXLFU-UHFFFAOYSA-N
SMILES[W].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration96.0 to 104.0 %