Chlorotrimethylsilane, 98%, AcroSeal™
Chlorotrimethylsilane, 98%, AcroSeal™
Chlorotrimethylsilane, 98%, AcroSeal™
Chlorotrimethylsilane, 98%, AcroSeal™
Chlorotrimethylsilane, 98%, AcroSeal™
Thermo Scientific Chemicals
Chlorotrimethylsilane, 98%, AcroSeal™
CAS: 75-77-4 | C3H9ClSi | 108.64 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA426431000100 mLAcroSeal™ Glass bottle
Catalog number FSA426431000
Price (MYR)
414.00
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
414.00
Add to cart
Specifications
Chemical Name or MaterialChlorotrimethylsilane
CAS75-77-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Harmful if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
Reacts violently with water.
Corrosive to the res
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
View more
Silylation reagent
RUO – Research Use Only