6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
6-Carboxyfluorescein, 96%
Thermo Scientific Chemicals
6-Carboxyfluorescein, 96%
CAS: 3301-79-9 | C21H12O7 | 376.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number FSA425801000
Price (MYR)
1,272.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,272.00
Add to cart
Chemical Identifiers
CAS109-69-3
IUPAC Name1-chlorobutane
Molecular FormulaC4H9Cl
InChI KeyVFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILESCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.7 %
Residue after evaporation=<2 ppm
UV1 cm cell vs HPLC-grade water
Appearance (Form)Liquid
Water=<0.020 %