Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 62-56-6 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA424542500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS938-09-0
IUPAC Name(2-chloroethanesulfonyl)benzene
Molecular FormulaC8H9ClO2S
InChI KeyNUJGORANFDSMOL-UHFFFAOYSA-N
SMILESClCCS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)50.0-57.0?C
Appearance (Color)White