Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number FSA424531000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-10-0
IUPAC Namepraseodymium
Molecular FormulaPr
InChI KeyPUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES[Pr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities excluding Ta0.5% max.