Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA424530250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS8068-28-8
IUPAC Namepentasodium [(3-{3-hydroxy-2-[2-(6-methyloctanamido)-4-[(sulfonatomethyl)amino]butanamido]butanamido}-3-{[3-(1-hydroxyethyl)-12,15-bis(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-6,9,18-tris({2-[(sulfonatomethyl)amino]ethyl})-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)amino]methanesulfonate
Molecular FormulaC58H105N16Na5O28S5
InChI KeyIQWHCHZFYPIVRV-UHFFFAOYNA-I
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].CCC(C)CCCCC(=O)NC(CCNCS([O-])(=O)=O)C(=O)NC(C(C)O)C(=O)NC(CCNCS([O-])(=O)=O)C(=O)NC1CCNC(=O)C(NC(=O)C(CCNCS([O-])(=O)=O)NC(=O)C(CCNCS([O-])(=O)=O)NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)C(CCNCS([O-])(=O)=O)NC1=O)C(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals≤ 0.003%
Loss on Drying≤ 5.0 %
pH6.5 to 8.5
Appearance (Color)White to off-white