5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA424511000
Price (MYR)
341.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
341.00
Add to cart
Chemical Identifiers
CAS2001-45-8
IUPAC Nametetraphenylphosphanium chloride
Molecular FormulaC24H20ClP
InChI KeyWAGFXJQAIZNSEQ-UHFFFAOYSA-M
SMILES[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)268-287?C
Indicator
RUO – Research Use Only