L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals
CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number FSA424440025
Price (MYR)
2,922.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
2,922.00
Add to cart
Chemical Identifiers
CAS51805-45-9
IUPAC Name3-[bis(2-carboxylatoethyl)phosphanyl]propanoate
Molecular FormulaC9H12O6P
InChI KeyPZBFGYYEXUXCOF-UHFFFAOYSA-K
SMILES[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream or pale yellow
Assay from Supplier's CofA≥97.5%