Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number FSA424335000
Price (MYR)
389.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
389.00
Add to cart
Chemical Identifiers
CAS3143-02-0
IUPAC Name(3-methyloxetan-3-yl)methanol
Molecular FormulaC5H10O2
InChI KeyNLQMSBJFLQPLIJ-UHFFFAOYSA-N
SMILESCC1(CO)COC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %