Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number FSA424330010
Price (MYR)
624.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
624.00
Add to cart
Chemical Identifiers
CAS92-52-4
IUPAC Name1,1'-biphenyl
Molecular FormulaC12H10
InChI KeyZUOUZKKEUPVFJK-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals or flakes
Melting point68°C to 72°C