Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic Bottle
Catalog number FSA424250025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS89-32-7
IUPAC Name5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
Molecular FormulaC10H2O6
InChI KeyANSXAPJVJOKRDJ-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Melting point283°C to 287°C
Infrared spectrumConforms
Titration excess NaOH>=98.5 %