Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number FSA424030010
Price (MYR)
2,802.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,802.00
Add to cart
Chemical Identifiers
CAS874-63-5
IUPAC Name1-methoxy-3,5-dimethylbenzene
Molecular FormulaC9H12O
InChI KeyJCHJBEZBHANKGA-UHFFFAOYSA-N
SMILESCOC1=CC(C)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5110-1.5140 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
FormLiquid
Oxidation reagent
RUO – Research Use Only