Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA423950010
Price (MYR)
299.00
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
299.00
Add to cart
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
CAS67-56-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.