Lithium perchlorate, 95+%, ACS reagent
Lithium perchlorate, 95+%, ACS reagent
Lithium perchlorate, 95+%, ACS reagent
Lithium perchlorate, 95+%, ACS reagent
Lithium perchlorate, 95+%, ACS reagent
Thermo Scientific Chemicals
Lithium perchlorate, 95+%, ACS reagent
CAS: 7791-03-9 | ClLiO4 | 106.39 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA423865000500 gPlastic bottle
Catalog number FSA423865000
Price (MYR)
1,805.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,805.00
Add to cart
Chemical Identifiers
CAS1892-57-5
IUPAC Name({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine
Molecular FormulaC8H17N3
InChI KeyLMDZBCPBFSXMTL-UHFFFAOYSA-N
SMILESCCN=C=NCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4600 to 1.4630 (20°C, 589 nm)
Suitable for battery materials development.
Oxidation reagent
RUO – Research Use Only