Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Catalog number FSA423830010
Price (MYR)
489.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
489.00
Add to cart
Chemical Identifiers
CAS6152-67-6
IUPAC Name4-(phenylamino)benzene-1-sulfonic acid sodium
Molecular FormulaC12H11NNaO3S
InChI KeyVMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to grey
CommentSensitivity as indicator P.T.