Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Plastic bottle
Catalog number FSA423795000
Price (MYR)
283.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
283.00
Add to cart
Chemical Identifiers
CAS5131-58-8
IUPAC Name4-nitrobenzene-1,3-diamine
Molecular FormulaC6H7N3O2
InChI KeyDPIZKMGPXNXSGL-UHFFFAOYSA-N
SMILESNC1=CC=C(C(N)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow to orange to red to brown
Melting Point (clear melt)152.0-164.0?C