Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA423790025
Price (MYR)
499.00
Add to cart
Quantity:
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
499.00
Add to cart
Chemical Identifiers
CAS2216-35-5
IUPAC Name2-bromononane
Molecular FormulaC9H19Br
InChI KeyJQEFZTLHNWFZDD-UHFFFAOYNA-N
SMILESCCCCCCCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4505-1.4555 @ 20?C
Appearance (Color)Clear colorless to pale yellow