Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic acid, 47.0 to 49.0%, BrH, CAS Number-10035-10-6, 7732-18-5, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, 126.0 deg.C to 128.0 deg.C, CHEBI:47266
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number FSA423780025
Price (MYR)
1,227.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,227.00
Add to cart
Chemical Identifiers
CAS50525-27-4
IUPAC Nameλ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride
Molecular FormulaC30H36Cl2N6O6Ru
InChI KeyWHELTKFSBJNBMQ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red
Elemental Analysis>=97.5 %
Appearance (Form)Crystalline powder or crystals
C-X bond formation reagent
RUO – Research Use Only