Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
Packaging:
Plastic bottle
Catalog number FSA423775000
Price (MYR)
932.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
932.00
Add to cart
Chemical Identifiers
CAS27298-99-3
IUPAC Name1-(4-chlorophenyl)ethan-1-amine
Molecular FormulaC8H10ClN
InChI KeyPINPOEWMCLFRRB-UHFFFAOYNA-N
SMILESCC(N)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to pale yellow
Enantiomeric excess>=97.5 %
Infrared spectrumConforms
Refractive index1.5400 to 1.5440 (20°C, 589 nm)
View more
Reduction reagent
RUO – Research Use Only