Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
CAS: 10034-93-2 | H4N2·H2SO4 | 130.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA423770500
Price (MYR)
318.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Specifications
Chemical Name or MaterialHydrazine sulfate
Melting Point254.0°C
CAS10034-93-2
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Reduction reagent
RUO – Research Use Only