Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent, Thermo Scientific Chemicals
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number FSA423750025
Price (MYR)
1,267.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,267.00
Add to cart
Chemical Identifiers
CAS1191-47-5
Molecular FormulaC8H18Mg
InChI KeyKJJBSBKRXUVBMX-UHFFFAOYSA-N
SMILES[Mg++].CCC[CH2-].CCC[CH2-]
Molecular Weight (g/mol)138.54
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to yellow
Titration10.1 to 12.2 %
Strength0.50 to 0.60 M
Appearance (Form)Solution