Diphenylamine-4-sulfonic acid, sodium salt, ACS reagent, Thermo Scientific Chemicals
Diphenylamine-4-sulfonic acid, sodium salt, ACS reagent, Thermo Scientific Chemicals
Diphenylamine-4-sulfonic acid, sodium salt, ACS reagent, Thermo Scientific Chemicals
Diphenylamine-4-sulfonic acid, sodium salt, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diphenylamine-4-sulfonic acid, sodium salt, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA423660050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13410-01-0
SpecificationsSpecification SheetSpecification Sheet
Identificationconfirmed
Appearance (Form)Crystalline powder
Assay>=97.5 %
Appearance (Color)White
Indicator
RUO – Research Use Only