N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA423640025
Price (MYR)
1,013.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,013.00
Add to cart
Chemical Identifiers
CAS82385-42-0
IUPAC Namesodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate
Molecular FormulaC7H4NNaO3S
InChI KeyOAZGZMKGRPRHJX-UHFFFAOYSA-M
SMILES[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<15 %
Readily carbonizable substancePasses test
Appearance (Color)Colorless to white
Titration with HClO499.0 to 101.0 % (on anhydrous substance)