N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8+%, ACS reagent, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA423640010
Price (MYR)
585.00
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
585.00
Add to cart
Chemical Identifiers
CAS3811-04-9
IUPAC Namepotassium chlorate
Molecular FormulaClKO3
InChI KeyVKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metals(as Pb) 5ppm
Assay (unspecified)99.0% KCLO₃ min
Impurity contentCl 0.001%
Impurity contentMg 0.002%
Impurity contentCa 0.002%