Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, 99.6%, Thermo Scientific Chemicals
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number FSA423560020
Price (MYR)
1,148.00
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
1,148.00
Add to cart
Chemical Identifiers
CAS98977-36-7
IUPAC Nametert-butyl 3-oxopiperidine-1-carboxylate
Molecular FormulaC10H17NO3
InChI KeyRIFXIGDBUBXKEI-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCCC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
Appearance (Form)Solid or liquid
Infrared spectrumConforms
GC>=96.0 %