Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur., Thermo Scientific Chemicals
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur., Thermo Scientific Chemicals
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur., Thermo Scientific Chemicals
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA423550010
Price (MYR)
516.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
516.00
Add to cart
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteStabilized
CAS67-66-3
Health Hazard 1Warning