Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content
Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content
Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content
Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content
Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content
Thermo Scientific Chemicals
Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content
CAS: 1704465-79-1 | C17H20ClN3 | 301.82 g/mol
Have Questions?
Catalog NumberQuantity
FSA4233400101 g
Catalog number FSA423340010
Price (MYR)
402.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
402.00
Add to cart
Chemical Identifiers
CAS108-98-5
IUPAC Namebenzenethiol
Molecular FormulaC6H6S
InChI KeyRMVRSNDYEFQCLF-UHFFFAOYSA-N
SMILESSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥99.0%
Refractive Index1.5880-1.5915 @ 20?C