Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
2.5 L
Catalog number FSA423300250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS20031-21-4
IUPAC Name(3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol
Molecular FormulaC8H14O5
InChI KeyJAUQZVBVVJJRKM-XZBKPIIZSA-N
SMILESCC1(C)O[C@H]2O[C@H](CO)[C@H](O)[C@H]2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Specific optical rotation-17° to -19° (20°C, 589 nm) (c=1, water)
Appearance (Color)White
Appearance (Form)Waxy solid or coarse crystals and/or chunks