Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA422750250
Price (MYR)
1,265.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,265.00
Add to cart
Chemical Identifiers
CAS675-14-9
IUPAC Nametrifluoro-1,3,5-triazine
Molecular FormulaC3F3N3
InChI KeyVMKJWLXVLHBJNK-UHFFFAOYSA-N
SMILESFC1=NC(F)=NC(F)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless or pale pink
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.3825-1.3865 @ 20°C (U.K. specification)
CommentPurchased in the U.K. and in other countries