Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc phthalocyanine, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA422750050
Price (MYR)
425.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
425.00
Add to cart
Chemical Identifiers
CAS108-86-1
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms (non-U.S. sourced material)
FormLiquid
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)Clear colorless to pale yellow
CommentMaterial sourced in the U.S. and in other countries