pH Indicator solution, universal (in isopropanol), pure
pH Indicator solution, universal (in isopropanol), pure
Thermo Scientific Chemicals
pH Indicator solution, universal (in isopropanol), pure
Have Questions?
Catalog NumberQuantity
FSA422435000500 mL
Catalog number FSA422435000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS67-63-0, 7732-18-5
Specifications
Chemical Name or MaterialpH Indicator solution, universal (in isopropanol)
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause drowsiness or dizziness.
CAS67-63-0
Health Hazard 3GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Indicator
RUO – Research Use Only