Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution, Thermo Scientific Chemicals
CAS: 14634-91-4 | C36H24FeN6O4S | 692.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA422251000
Price (MYR)
497.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
497.00
Add to cart
Specifications
Chemical Name or MaterialTris(1, 10-phenanthroline)iron(II) sulfate
Name Note0.025M Aqueous Solution
CAS14634-91-4
ColorRed
Density1.0060g/mL
View more
Indicator
RUO – Research Use Only