2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA421985000
Price (MYR)
352.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
352.00
Add to cart
Chemical Identifiers
CAS95838-16-7
IUPAC Nameλ⁴-cerium(4+) disulfate
Molecular FormulaCeO8S2
InChI KeyVZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities1% max.
For UV/VIS spectroscopy
RUO – Research Use Only