2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, Reagent ACS, Spectro Grade, +99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass Bottle
Catalog number FSA421980025
Price (MYR)
828.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
828.00
Add to cart
Specifications
Chemical Name or Material2, 2, 4-Trimethylpentane
Melting Point-107.0°C
Absorbance0.01 max. at 250nm, 0.01 max. at 400nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 1 max. at 210nm
CAS540-84-1
CAS Min %99.0
View more
For UV/VIS spectroscopy
RUO – Research Use Only