1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
1,1,2-Trimethyl-1H-benz[e]indole, 96%
Alfa Aesar
1,1,2-Trimethyl-1H-benz[e]indole, 96%
CAS: 41532-84-7 | C15H15N | 209.29 g/mol
Have Questions?
Catalog number FSA421900250
Price (MYR)
602.00
Add to cart
Request bulk or custom format
Price (MYR)
602.00
Add to cart
Chemical Identifiers
CAS578-54-1
IUPAC Name2-ethylaniline
Molecular FormulaC8H11N
InChI KeyMLPVBIWIRCKMJV-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to red to red-brown or pink
FormLiquid
Refractive Index1.5590-1.5620 @ 20?C
Assay (GC)≥97.5%