Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
CAS: 87-90-1 | C3Cl3N3O3 | 232.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass Bottle
Catalog number FSA421540010
Price (MYR)
532.00
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
532.00
Add to cart
Chemical Identifiers
CAS68-34-8
IUPAC Name4-methyl-N-phenylbenzene-1-sulfonamide
Molecular FormulaC13H13NO2S
InChI KeyVLVCWODDMDGANW-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
C-X bond formation reagent
RUO – Research Use Only