2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals
CAS: 75-80-9 | C2H3Br3O | 282.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA421430100
Price (MYR)
419.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
419.00
Add to cart
Chemical Identifiers
CAS65-49-6
IUPAC Name4-amino-2-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyWUBBRNOQWQTFEX-UHFFFAOYSA-N
SMILESNC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=98.5 %
Appearance (Color)Beige