p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA421211000100 gPlastic bottle
Catalog number FSA421211000
Price (MYR)
252.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
252.00
Add to cart
Chemical Identifiers
CAS15878-77-0
IUPAC Namepraseodymium(3+) hexahydrate trinitrate
Molecular FormulaH12N3O15Pr
InChI KeyLXXCECZPOWZKLC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale green
FormCrystalline
Assay (special)38.3-40.0% Pr₆O{11}
Total Rare Earth Oxide Impurities≤0.1% (1000 ppm)