o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 99%, Thermo Scientific Chemicals
CAS: 612-82-8 | C14H16N2·2HCl | 285.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass Bottle
Catalog number FSA421131000
Price (MYR)
949.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
949.00
Add to cart
Specifications
Chemical Name or Materialo-Tolidine dihydrochloride
CAS612-82-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.