DL-α-Tocopheryl acetate, 98%
DL-α-Tocopheryl acetate, 98%
DL-α-Tocopheryl acetate, 98%
DL-α-Tocopheryl acetate, 98%
Thermo Scientific Chemicals
DL-α-Tocopheryl acetate, 98%
CAS: 7695-91-2 | C31H52O3 | 472.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA421070010
Price (MYR)
267.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
267.00
Add to cart
Chemical Identifiers
CAS614-45-9
IUPAC Nametert-butyl benzenecarboperoxoate
Molecular FormulaC11H14O3
InChI KeyGJBRNHKUVLOCEB-UHFFFAOYSA-N
SMILESCC(C)(C)OOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Assay (Titration)≥97.5 to ≤102.5%
Refractive Index1.4970-1.5010 @ 20?C
Appearance (Color)Clear, colorless to yellow